Assay Details
| Gene Symbol: Gbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233425.2 | XP_006233487.1 | 7 - 8 | 1306 | 90 | |
| XM_008761538.1 | XP_008759760.1 | 8 - 9 | 1501 | 90 | ||
More Information
Set Membership: |
|

Gbp1
Genomic Map