Assay Details
| Gene Symbol: Gbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233425.2 | XP_006233487.1 | 9 - 10 | 1621 | 104 | |
| XM_008761538.1 | XP_008759760.1 | 10 - 11 | 1816 | 104 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gbp1
Genomic Map