Assay Details
| Gene Symbol: Tspyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191618.2 | NP_001178547.1 | 1 - 2 | 909 | 87 | |
| XM_006257038.2 | XP_006257100.1 | 1 - 2 | 1060 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tspyl2
Genomic Map