Assay Details
| Gene Symbol: Iqsec2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277386.1 | NP_001264315.1 | 1970 | 59 | ||
| NM_001277425.1 | NP_001264354.1 | 2735 | 59 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Iqsec2
Genomic Map