Assay Details
| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109088.1 | NP_001102558.1 | 17 - 18 | 1738 | 65 | |
| XM_006250963.1 | XP_006251025.1 | 17 - 18 | 1808 | 65 | ||
| XM_006250965.2 | XP_006251027.1 | 17 - 18 | 2208 | 65 | ||
| GenBank mRNA | FQ222791.1 | - | 15 - 16 | 2031 | 65 | |
| FQ225520.1 | - | 1 - 2 | 79 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of transcription from RNA polymerase II promoter
cation transmembrane transport
regulation of nucleic acid-templated transcription
chromatin binding
cation transmembrane transporter activity
ligand-dependent nuclear receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
positive regulation of transcription from RNA polymerase II promoter
cation transmembrane transport
regulation of nucleic acid-templated transcription
chromatin binding
cation transmembrane transporter activity
ligand-dependent nuclear receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P234087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76499 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P234087 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P234087 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P234087 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P234087 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P603500 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36217 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63864 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10776686 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776682 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776695 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776687 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776683 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776692 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776684 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776696 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776698 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776688 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776693 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776690 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776694 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776697 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776691 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776689 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776685 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10776681 | - | - |
| Rat Genome 230 2.0 Array | 1375514_at | - | - |
| Rat Genome 230 2.0 Array | 1395340_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388982_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375514_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395340_at | - | - |
| Rat Genome 230 2.0 Array | 1388982_at | - | - |
| Affymetrix HT Rat Focus Array | 1395340_at | - | - |
| Affymetrix HT Rat Focus Array | 1395340_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375514_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375514_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400018441.rn.1 | TC1400002474.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388982_PM_at | - | - |
| Rat Expression Set 230 | 1375514_PM_at | - | - |
| Rat Expression Set 230 | 1388982_at | - | - |
| Rat Expression Set 230 | 1375514_at | - | - |
| Rat Expression Set 230 | 1395340_at | - | - |
| Rat Expression Set 230 | 1395340_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776681 | - | - |
| Rat Genome U34 Set | rc_AA942763_at | - | - |
| Rat Genome U34 Set | rc_AI012457_at | - | - |
| Rat Genome U34 Set | rc_AI103978_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21589912 | - | - |

Slc30a9
Genomic Map