Assay Details
| Gene Symbol: Vom2r47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099506.1 | NP_001092976.1 | 2 - 3 | 1137 | 96 | |
| XM_006232462.1 | XP_006232524.1 | 3 - 4 | 1242 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vom2r47
Genomic Map