Assay Details
| Gene Symbol: NEWGENE_1595998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767890.1 | XP_008766112.1 | 3 - 4 | 1266 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEWGENE_1595998| Gene Symbol: NEWGENE_1595998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767890.1 | XP_008766112.1 | 3 - 4 | 1266 | 55 | |
Set Membership: |
|