Assay Details
| Gene Symbol: Prrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276470.1 | NP_001263399.1 | 1 - 2 | 133 | 165 | |
| XM_006230308.2 | XP_006230370.1 | 1 - 2 | 212 | 165 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001276470.1, XM_006230308.2 transcripts.
Set Membership: |
|

Prrt2
Genomic Map