Assay Details
| Gene Symbol: Cs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130755.1 | NP_570111.1 | 1 - 2 | 112 | 95 | |
| GenBank mRNA | AF461496.1 | - | 1 - 2 | 112 | 95 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF461496.1,NM_130755.1 transcripts.
Set Membership: |
|

Cs
Genomic Map