Assay Details
| Gene Symbol: Cs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130755.1 | NP_570111.1 | 3 - 4 | 276 | 154 | |
| GenBank mRNA | AF461496.1 | - | 3 - 4 | 276 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cs
Genomic Map