Assay Details
| Gene Symbol: Prl8a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021580.1 | NP_067591.1 | 1 - 2 | 96 | 153 | |
| GenBank mRNA | AB009889.1 | - | 1 - 2 | 96 | 153 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB009889.1, NM_021580.1 transcripts.
Set Membership: |
|

Prl8a4
Genomic Map