Assay Details
| Gene Symbol: LOC100909582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249676.2 | XP_006249738.1 | 3 - 4 | 883 | 62 | |
| XM_006249677.2 | XP_006249739.1 | 3 - 4 | 880 | 62 | ||
| XM_006249679.2 | XP_006249741.1 | 2 - 3 | 455 | 62 | ||
| XM_008769509.1 | XP_008767731.1 | 4 - 5 | 1031 | 62 | ||
| Gene Symbol: Mgat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023095.1 | NP_075583.1 | 3 - 4 | 531 | 62 | |
| GenBank mRNA | L14284.1 | - | 3 - 4 | 531 | 62 | |
More Information
Set Membership: |
|

LOC100909582
Genomic Map