Assay Details
| Gene Symbol: Napepld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199381.1 | NP_955413.1 | 1 - 2 | 291 | 69 | |
| GenBank mRNA | AB112351.1 | - | 1 - 2 | 291 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Napepld
Genomic Map