Assay Details
| Gene Symbol: Rnase6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206815.1 | NP_996538.1 | 1 | 419 | 61 | |
| XM_006251871.2 | XP_006251933.1 | 2 | 488 | 61 | ||
| XM_006251872.2 | XP_006251934.1 | 2 | 787 | 61 | ||
| XM_008770517.1 | XP_008768739.1 | 3 | 632 | 61 | ||
| XM_008770518.1 | XP_008768740.1 | 3 | 606 | 61 | ||
| XM_008770519.1 | XP_008768741.1 | 2 | 506 | 61 | ||
| XM_008770520.1 | XP_008768742.1 | 2 | 485 | 61 | ||
| Gene Symbol: LOC103694852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773787.1 | XP_008772009.1 | 2 | 488 | 61 | |
| XM_008773788.1 | XP_008772010.1 | 2 | 787 | 61 | ||
| XM_008773789.1 | XP_008772011.1 | 3 | 635 | 61 | ||
| XM_008773790.1 | XP_008772012.1 | 3 | 609 | 61 | ||
| XM_008773791.1 | XP_008772013.1 | 2 | 509 | 61 | ||
| XM_008773792.1 | XP_008772014.1 | 2 | 485 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnase6
Genomic Map