Assay Details
| Gene Symbol: Olr1701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001114.1 | NP_001001114.1 | 1 | 688 | 129 | |
| XM_006255861.2 | XP_006255923.1 | 2 | 879 | 129 | ||
| XM_006255863.2 | XP_006255925.1 | 2 | 848 | 129 | ||
| XM_006255865.2 | XP_006255927.1 | 1 | 694 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Olr1701
Genomic Map