Assay Details
| Gene Symbol: Vcsa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012684.4 | NP_036816.4 | 2 - 3 | 117 | 134 | |
| GenBank mRNA | A07543.1 | - | 2 - 3 | 124 | 134 | |
| CB769246.1 | - | 2 - 3 | 129 | 134 | ||
| M63112.1 | - | 2 - 3 | 124 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P001104 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P468348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154077 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P468348 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P001104 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P001104 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P468348 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P468348 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P001104 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15401 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P468348 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58928 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18529 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28936 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95225 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17145 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25482 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10776115 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776108 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776118 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10776116 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10776104 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10776114 | - | - |
| Rat Genome 230 2.0 Array | 1369114_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387463_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387463_at | - | - |
| Rat Genome 230 2.0 Array | 1369114_at | - | - |
| Affymetrix HT Rat Focus Array | 1387463_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387463_at | - | - |
| Affymetrix HT Rat Focus Array | 1369114_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369114_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400011404.rn.1 | TC1400001543.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400011423.rn.1 | TC1400001543.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400011406.rn.1 | TC1400001543.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369114_PM_at | - | - |
| Rat Expression Set 230 | 1387463_at | - | - |
| Rat Expression Set 230 | 1369114_at | - | - |
| Rat Expression Set 230 | 1387463_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776104 | - | - |
| Rat Genome U34 Set | X84997mRNA#5_at | - | - |
| Rat Genome U34 Set | A07543cds_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349405 | - | - |
| RatRef-12 Expression BeadChip | ILMN_60485 | - | - |

Vcsa1
Genomic Map