Assay Details
| Gene Symbol: Ns5atp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207607.1 | NP_997490.1 | 1 | 443 | 120 | |
| GenBank mRNA | AY533136.1 | - | 1 | 443 | 120 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ns5atp4
Genomic Map