Assay Details
| Gene Symbol: Gapt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109491.1 | NP_001102961.1 | 2 | 1657 | 143 | |
| XM_006231916.2 | XP_006231978.1 | 2 | 1733 | 143 | ||
| XM_006231917.2 | XP_006231979.1 | 2 | 1693 | 143 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gapt
Genomic Map