Assay Details
| Gene Symbol: LOC100912008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233726.2 | XP_006233788.1 | 9 | 95 | ||
| XM_006233727.2 | XP_006233789.1 | 95 | ||||
| XM_006233728.2 | XP_006233790.1 | 95 | ||||
More Information
Set Membership: |
|

LOC100912008
Genomic Map