Assay Details
| Gene Symbol: Bdkrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030851.1 | NP_110478.1 | 2 | 1029 | 92 | |
| XM_008764861.1 | XP_008763083.1 | 1 | 907 | 92 | ||
| GenBank mRNA | AJ132230.2 | - | 2 | 1029 | 92 | |
| U66107.2 | - | 2 | 1038 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bdkrb1
Genomic Map