Assay Details
| Gene Symbol: Zfp771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759932.1 | XP_008758154.1 | 3 | 2001 | 145 | |
| XM_008759933.1 | XP_008758155.1 | 3 | 1122 | 145 | ||
| GenBank mRNA | FQ210015.1 | - | 3 | 1112 | 145 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp771
Genomic Map