Assay Details
| Gene Symbol: Zfp691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107968.2 | NP_001101438.1 | 1 | 1377 | 124 | |
| XM_006238757.2 | XP_006238819.1 | 2 | 2595 | 124 | ||
| XM_008764010.1 | XP_008762232.1 | 2 | 1424 | 124 | ||
| XM_008764011.1 | XP_008762233.1 | 2 | 1876 | 124 | ||
| GenBank mRNA | BC166565.1 | - | 1 | 1377 | 7316740 | 124 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp691
Genomic Map