Assay Details
| Gene Symbol: Dand5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001064979.3 | XP_001064979.1 | 2 | 3222 | 108 | |
| XM_006255337.2 | XP_006255399.1 | 3 | 2756 | 108 | ||
| XM_006255338.2 | XP_006255400.1 | 3 | 2811 | 108 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dand5
Genomic Map