Assay Details
| Gene Symbol: Adss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105975.1 | NP_001099445.1 | 13 | 2436 | 159 | |
| GenBank mRNA | FQ231775.1 | - | 1 | 481 | 159 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide biosynthetic process
AMP biosynthetic process
aspartate metabolic process
response to purine-containing compound
'de novo' AMP biosynthetic process
IMP metabolic process
response to ammonium ion
cellular response to electrical stimulus
magnesium ion binding
adenylosuccinate synthase activity
GTP binding
purine nucleotide biosynthetic process
AMP biosynthetic process
aspartate metabolic process
response to purine-containing compound
'de novo' AMP biosynthetic process
IMP metabolic process
response to ammonium ion
cellular response to electrical stimulus
magnesium ion binding
adenylosuccinate synthase activity
GTP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32746 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14869 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14869 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14869 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P207721 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P534211 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14869 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46176 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25299 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10770211 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770215 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770220 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770212 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770217 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770214 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770210 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770213 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770221 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770218 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770209 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770216 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770219 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10770208 | - | - |
| Rat Genome 230 2.0 Array | 1399050_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375587_at | - | - |
| Rat Genome 230 2.0 Array | 1375587_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1399050_at | - | - |
| Affymetrix HT Rat Focus Array | 1399050_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1399050_at | - | - |
| Affymetrix HT Rat Focus Array | 1375587_at | - | - |
| Affymetrix HT Rat Focus Array | 1375587_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300014436.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014437.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014431.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014441.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014433.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014439.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014430.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014435.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014438.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014440.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014442.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014434.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014445.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014432.rn.1 | TC1300002031.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300014443.rn.1 | TC1300002031.rn.1 | RTA TC |
| Rat Expression Set 230 | 1399050_at | - | - |
| Rat Expression Set 230 | 1375587_at | - | - |
| Rat Expression Set 230 | 1399050_PM_at | - | - |
| Rat Expression Set 230 | 1375587_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770208 | - | - |
| Rat Genome U34 Set | rc_AA963094_at | - | - |
| Rat Genome U34 Set | rc_AI230617_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22328333 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355936 | - | - |

Adss
Genomic Map