Assay Details
| Gene Symbol: Acot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138907.2 | NP_620262.2 | 3 | 1171 | 102 | |
| XM_006240320.2 | XP_006240382.1 | 3 | 1255 | 102 | ||
| GenBank mRNA | BC072540.1 | - | 3 | 1171 | 7104319 | 102 |
| Y09333.1 | - | 3 | 1112 | 102 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Acot2
Genomic Map