Assay Details
| Gene Symbol: Ston2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240406.2 | XP_006240468.1 | 7 | 4379 | 117 | |
| XM_006240407.2 | XP_006240469.1 | 7 | 4117 | 117 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ston2
Genomic Map