Assay Details
| Gene Symbol: Pxmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031587.1 | NP_113775.1 | 3 | 372 | 133 | |
| GenBank mRNA | X70223.1 | - | 3 | 372 | 133 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pxmp2
Genomic Map