Assay Details
| Gene Symbol: Oas1i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009680.1 | NP_001009680.1 | 1 | 73 | 110 | |
| GenBank mRNA | AY196697.1 | - | 1 | 73 | 110 | |
| BC160819.1 | - | 1 | 47 | 7382468 | 110 | |
| Gene Symbol: LOC100910735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751197.2 | XP_003751245.1 | 93 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oas1i
Genomic Map