Assay Details
| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108341.1 | NP_001101811.1 | 28 | 4756 | 113 | |
| XM_006249565.2 | XP_006249627.1 | 28 | 5080 | 113 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
Ras protein signal transduction
axonogenesis
protein localization
regulation of autophagy
positive regulation of autophagy
positive regulation of macroautophagy
cerebellar granule cell differentiation
radial glia guided migration of cerebellar granule cell
neuron projection regeneration
neuron projection development
receptor internalization
cellular response to nutrient levels
response to starvation
protein autophosphorylation
negative regulation of collateral sprouting
axon extension
regulation of neurotrophin TRK receptor signaling pathway
molecular_function
protein kinase activity
protein serine/threonine kinase activity
ATP binding
Rab GTPase binding
protein kinase binding
protein complex binding
autophagosome assembly
Ras protein signal transduction
axonogenesis
protein localization
regulation of autophagy
positive regulation of autophagy
positive regulation of macroautophagy
cerebellar granule cell differentiation
radial glia guided migration of cerebellar granule cell
neuron projection regeneration
neuron projection development
receptor internalization
cellular response to nutrient levels
response to starvation
protein autophosphorylation
negative regulation of collateral sprouting
axon extension
regulation of neurotrophin TRK receptor signaling pathway
molecular_function
protein kinase activity
protein serine/threonine kinase activity
ATP binding
Rab GTPase binding
protein kinase binding
protein complex binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P039387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131708 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039387 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039387 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P039387 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P503673 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9846 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100382 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10759274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759252 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759264 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759255 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759268 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759263 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759262 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759265 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759266 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759260 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759256 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759257 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759269 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759253 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759254 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759250 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759259 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759261 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759267 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759258 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759251 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759271 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10759248 | - | - |
| Rat Genome 230 2.0 Array | 1382402_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375869_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375869_at | - | - |
| Rat Genome 230 2.0 Array | 1382402_at | - | - |
| Affymetrix HT Rat Focus Array | 1382402_at | - | - |
| Affymetrix HT Rat Focus Array | 1375869_at | - | - |
| Affymetrix HT Rat Focus Array | 1382402_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375869_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1200007600.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007602.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007605.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007615.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007620.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007612.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007616.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007617.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007596.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007603.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007610.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007618.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007622.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007608.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007593.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007599.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007606.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007607.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007609.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007614.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007594.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007595.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007601.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007604.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007613.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007623.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007597.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007598.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007611.rn.1 | TC1200000868.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200007619.rn.1 | TC1200000868.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375869_PM_at | - | - |
| Rat Expression Set 230 | 1375869_at | - | - |
| Rat Expression Set 230 | 1382402_at | - | - |
| Rat Expression Set 230 | 1382402_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759248 | - | - |
| Rat Genome U34 Set | rc_AA849717_at | - | - |
| Rat Genome U34 Set | rc_AI230479_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21137543 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362810 | - | - |

Ulk1
Genomic Map