Assay Details
| Gene Symbol: Dse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108933.1 | NP_001102403.1 | 6 | 3745 | 109 | |
| XM_006256382.2 | XP_006256444.1 | 6 | 3500 | 109 | ||
| XM_008772900.1 | XP_008771122.1 | 6 | 3577 | 109 | ||
| XM_008772901.1 | XP_008771123.1 | 8 | 4079 | 109 | ||
| GenBank mRNA | BC168891.1 | - | 8 | 4013 | 7110121 | 109 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P579594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88128 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P579594 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P579594 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P579594 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P579594 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P328573 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3329 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55380 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10830260 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830256 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830257 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830258 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830259 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830255 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10830253 | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000012218.rn.1 | TC2000001494.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012212.rn.1 | TC2000001494.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012213.rn.1 | TC2000001494.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012209.rn.1 | TC2000001494.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012210.rn.1 | TC2000001494.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012223.rn.1 | TC2000001494.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012217.rn.1 | TC2000001494.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012211.rn.1 | TC2000001494.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012221.rn.1 | TC2000001494.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012224.rn.1 | TC2000001494.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012220.rn.1 | TC2000001494.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10830253 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20786415 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356076 | - | - |

Dse
Genomic Map