Assay Details
| Gene Symbol: Als2cr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014101.1 | NP_001014123.1 | 1 | 321 | 117 | |
| GenBank mRNA | BC079241.1 | - | 1 | 321 | 7132659 | 117 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC079241.1, NM_001014101.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P164138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95843 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P164138 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P164138 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P164138 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P164138 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103585 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85423 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25413 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57334 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68142 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10928292 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928295 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928300 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928305 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928303 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928306 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928301 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928302 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928296 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928299 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928304 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928297 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928298 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928291 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10928290 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900016778.rn.1 | TC0900002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016788.rn.1 | TC0900002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016777.rn.1 | TC0900002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016784.rn.1 | TC0900002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016786.rn.1 | TC0900002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016787.rn.1 | TC0900002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016776.rn.1 | TC0900002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016779.rn.1 | TC0900002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016785.rn.1 | TC0900002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016782.rn.1 | TC0900002034.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10928290 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21199144 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350975 | - | - |

Als2cr12
Genomic Map