Assay Details
| Gene Symbol: Klf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108800.1 | NP_001102270.1 | 3 | 2854 | 133 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P163987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146884 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P163987 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P163987 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P163987 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P439473 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P439475 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84001 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10928526 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928525 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928527 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10928524 | - | - |
| Rat Genome 230 2.0 Array | 1384497_at | - | - |
| Rat Genome 230 2.0 Array | 1384497_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391471_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380363_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391471_at | - | - |
| Rat Genome 230 2.0 Array | 1377618_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377618_at | - | - |
| Rat Genome 230 2.0 Array | 1380363_at | - | - |
| Affymetrix HT Rat Focus Array | 1377618_at | - | - |
| Affymetrix HT Rat Focus Array | 1391471_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377618_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380363_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391471_at | - | - |
| Affymetrix HT Rat Focus Array | 1384497_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380363_at | - | - |
| Affymetrix HT Rat Focus Array | 1384497_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900017357.rn.1 | TC0900002107.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900017365.rn.1 | TC0900002107.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900017358.rn.1 | TC0900002107.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900017364.rn.1 | TC0900002107.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900017356.rn.1 | TC0900002107.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900017359.rn.1 | TC0900002107.rn.1 | RTA TC |
| Rat Expression Set 230 | 1377618_PM_at | - | - |
| Rat Expression Set 230 | 1384497_at | - | - |
| Rat Expression Set 230 | 1391471_at | - | - |
| Rat Expression Set 230 | 1384497_PM_at | - | - |
| Rat Expression Set 230 | 1380363_PM_at | - | - |
| Rat Expression Set 230 | 1377618_at | - | - |
| Rat Expression Set 230 | 1380363_at | - | - |
| Rat Expression Set 230 | 1391471_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928524 | - | - |
| Rat Genome U34 Set | rc_AI229204_at | - | - |
| Rat Genome U34 Set | rc_AI101189_at | - | - |
| Rat Genome U34 Set | rc_AI234733_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21552180 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349559 | - | - |

Klf7
Genomic Map