Assay Details
| Gene Symbol: Kcnj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766044.1 | XP_008764266.1 | 3 | 2842 | 141 | |
| XM_008766045.1 | XP_008764267.1 | 3 | 3532 | 141 | ||
| XM_008766046.1 | XP_008764268.1 | 2 | 2720 | 141 | ||
| XM_008766047.1 | XP_008764269.1 | 2 | 2833 | 141 | ||
| XM_008766048.1 | XP_008764270.1 | 2 | 3410 | 141 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kcnj1
Genomic Map