Assay Details
| Gene Symbol: Bmper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135799.1 | NP_001129271.1 | 15 | 3721 | 90 | |
| XM_006242715.2 | XP_006242777.1 | 13 | 4675 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
ureteric bud development
blood vessel endothelial cell proliferation involved in sprouting angiogenesis
regulation of endothelial cell migration
negative regulation of BMP signaling pathway
endothelial cell activation
endothelial cell differentiation
inner ear development
regulation of pathway-restricted SMAD protein phosphorylation
positive regulation of ERK1 and ERK2 cascade
ureteric bud development
blood vessel endothelial cell proliferation involved in sprouting angiogenesis
regulation of endothelial cell migration
negative regulation of BMP signaling pathway
endothelial cell activation
endothelial cell differentiation
inner ear development
regulation of pathway-restricted SMAD protein phosphorylation
positive regulation of ERK1 and ERK2 cascade
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P151779 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P151774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108114 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151779 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151774 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151779 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151774 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P151779 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P151774 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P220511 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10908650 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908653 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908651 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908648 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908654 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908647 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908646 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908649 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908652 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10908645 | - | - |
| Rat Genome 230 2.0 Array | 1391345_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391345_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800001761.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001771.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001776.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001781.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001762.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001766.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001767.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001768.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001775.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001778.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001774.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001780.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001763.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001770.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001772.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001773.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001779.rn.1 | TC0800000264.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800001777.rn.1 | TC0800000264.rn.1 | RTA TC |
| Rat Expression Set 230 | 1391345_at | - | - |
| Rat Expression Set 230 | 1391345_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21325396 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353604 | - | - |

Bmper
Genomic Map