Assay Details
| Gene Symbol: Sub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232043.2 | XP_006232105.1 | 5 | 3097 | 93 | |
| GenBank mRNA | FQ211681.1 | - | 5 | 3087 | 93 | |
| FQ211748.1 | - | 5 | 3087 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sub1
Genomic Map