Assay Details
| Gene Symbol: Efhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109310.1 | NP_001102780.1 | 4 | 1741 | 150 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P17795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126841 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P17795 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P17795 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P17795 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17795 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P320292 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 32584 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10925016 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925018 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925015 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925017 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10925014 | - | - |
| Rat Genome 230 2.0 Array | 1376770_at | - | - |
| Rat Genome 230 2.0 Array | 1376770_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376770_at | - | - |
| Affymetrix HT Rat Focus Array | 1376770_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900009124.rn.1 | TC0900001077.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376770_at | - | - |
| Rat Expression Set 230 | 1376770_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925014 | - | - |
| Rat Genome U34 Set | rc_AI012471_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21921529 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371509 | - | - |

Efhd1
Genomic Map