Assay Details
| Gene Symbol: Repin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006236423.2 | XP_006236485.1 | 1 | 850 | 107 | |
| XM_006236424.2 | XP_006236486.1 | 1 | 850 | 107 | ||
| XM_006236427.2 | XP_006236489.1 | 2 | 971 | 107 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Repin1
Genomic Map