Assay Details
| Gene Symbol: Chst7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207600.2 | NP_997483.1 | 1 | 1246 | 115 | |
| GenBank mRNA | AY216524.1 | - | 1 | 1135 | 115 | |
| BC111079.1 | - | 1 | 1246 | 7387313 | 115 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chst7
Genomic Map