Assay Details
| Gene Symbol: P2ry13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002853.1 | NP_001002853.1 | 1 | 955 | 159 | |
| XM_008760991.1 | XP_008759213.1 | 2 | 1261 | 159 | ||
| GenBank mRNA | AY639875.1 | - | 1 | 955 | 159 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

P2ry13
Genomic Map