Assay Details
| Gene Symbol: Krt32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008818.1 | NP_001008818.1 | 2 - 3 | 570 | 142 | |
| XM_006247394.2 | XP_006247456.1 | 2 - 3 | 594 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt32
Genomic Map