Assay Details
| Gene Symbol: RT1-Ha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008848.2 | NP_001008848.1 | 2 - 3 | 379 | 168 | |
| XM_006255916.2 | XP_006255978.2 | 2 - 3 | 461 | 168 | ||
| GenBank mRNA | BC099182.1 | - | 1 - 2 | 581 | 7383188 | 168 |
| Gene Symbol: LOC100911800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751931.3 | XP_003751979.3 | 2 - 3 | 461 | 168 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

RT1-Ha
Genomic Map