Assay Details
| Gene Symbol: Yod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008889.1 | NP_001008889.1 | 1 - 2 | 210 | 133 | |
| GenBank mRNA | BC107904.1 | - | 1 - 2 | 703 | 7454897 | 133 |
More Information
Updated Mapping Information
Set Membership: |
|

Yod1
Genomic Map