Assay Details
| Gene Symbol: Nipsnap3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009422.1 | NP_001009422.1 | 3 - 4 | 474 | 153 | |
| XM_006238164.2 | XP_006238226.1 | 3 - 4 | 529 | 153 | ||
| GenBank mRNA | BC087643.1 | - | 3 - 4 | 474 | 7307943 | 153 |
| FQ219476.1 | - | 3 - 4 | 526 | 153 | ||
| FQ221422.1 | - | 3 - 4 | 443 | 153 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nipsnap3b
Genomic Map