Assay Details
| Gene Symbol: Oas1k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009489.1 | NP_001009489.1 | 1 - 2 | 238 | 150 | |
| XM_006249400.2 | XP_006249462.1 | 1 - 2 | 214 | 150 | ||
| XM_006249401.2 | XP_006249463.1 | 1 - 2 | 214 | 150 | ||
| GenBank mRNA | AY196696.1 | - | 1 - 2 | 238 | 150 | |
| BC088437.1 | - | 1 - 2 | 187 | 7125462 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oas1k
Genomic Map