Assay Details
| Gene Symbol: Oas1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009491.1 | NP_001009491.1 | 3 - 4 | 697 | 136 | |
| XM_008769247.1 | XP_008767469.1 | 3 - 4 | 774 | 136 | ||
| GenBank mRNA | AY196701.1 | - | 3 - 4 | 697 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oas1h
Genomic Map