Assay Details
| Gene Symbol: Bin2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009524.1 | NP_001009524.1 | 13 - 14 | 1469 | 136 | |
| XM_006242737.2 | XP_006242799.1 | 13 - 14 | 1347 | 136 | ||
| GenBank mRNA | AF299297.1 | - | 13 - 14 | 1469 | 136 | |
| AF299298.1 | - | 13 - 14 | 1323 | 136 | ||
| AF414431.1 | - | 13 - 14 | 1323 | 136 | ||
| AY040870.1 | - | 13 - 14 | 1469 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bin2a
Genomic Map