Assay Details
| Gene Symbol: Spsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009660.1 | NP_001009660.1 | 2 - 3 | 873 | 173 | |
| XM_006237336.2 | XP_006237398.1 | 2 - 3 | 1362 | 173 | ||
| XM_006237337.2 | XP_006237399.1 | 2 - 3 | 909 | 173 | ||
| GenBank mRNA | BC088189.1 | - | 2 - 3 | 873 | 7371446 | 173 |
| Gene Symbol: LOC100911548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750655.3 | XP_003750703.1 | 2 - 3 | 985 | 173 | |
| XM_006244625.2 | XP_006244687.1 | 2 - 3 | 1358 | 173 | ||
| XM_006244626.2 | XP_006244688.1 | 2 - 3 | 907 | 173 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spsb2
Genomic Map