Assay Details
| Gene Symbol: RGD1311458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009678.1 | NP_001009678.1 | 7 - 8 | 1022 | 128 | |
| GenBank mRNA | BC088456.1 | - | 7 - 8 | 1022 | 7127273 | 128 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311458
Genomic Map