Assay Details
| Gene Symbol: Oas1i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009680.1 | NP_001009680.1 | 4 - 5 | 917 | 143 | |
| GenBank mRNA | AY196697.1 | - | 4 - 5 | 917 | 143 | |
| BC160819.1 | - | 4 - 5 | 891 | 7382468 | 143 | |
| Gene Symbol: LOC100910735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751197.2 | XP_003751245.1 | 937 | 143 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Oas1i
Genomic Map