Assay Details
| Gene Symbol: Mad2l1bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009699.1 | NP_001009699.1 | 1 - 2 | 62 | 71 | |
| GenBank mRNA | BC088268.1 | - | 1 - 2 | 62 | 7379293 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Mad2l1bp
Genomic Map